Gene Transcripts

Gene Symbol
Det1
Entrez Gene ID
76375
Gene Name
de-etiolated homolog 1 (Arabidopsis)
Gene Aliases
2610034H20Rik
Chromosome Location
Chr. 7: 78822959 - 78847382 on Build GRCm38
UniGene ID
Mm.250541
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029585.3 NP_083861.1 2 784
GenBank mRNA AK011666.1 - 2 810
BC031930.1 - 2 784 4018106

View Full Product Details