Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000398.6 | NP_000389.1 | 8 | 761 | |
NM_001129819.2 | NP_001123291.1 | 8 | 1005 | ||
NM_001171660.1 | NP_001165131.1 | 8 | 901 | ||
NM_001171661.1 | NP_001165132.1 | 9 | 821 | ||
NM_007326.4 | NP_015565.1 | 8 | 760 | ||
GenBank mRNA | AF361370.1 | - | 8 | 696 | |
AJ010116.1 | - | 8 | 674 | ||
AJ010117.1 | - | 8 | 674 | ||
AJ010118.1 | - | 8 | 674 | ||
AJ310899.1 | - | 5 | 378 | ||
AJ310900.1 | - | 5 | 378 | ||
AK124528.1 | - | 5 | 2811 | ||
AK302204.1 | - | 8 | 900 | ||
BC004821.1 | - | 8 | 705 | 3450773 | |
BM564146.1 | - | 9 | 810 | ||
BT009821.1 | - | 8 | 677 | ||
BX334220.2 | - | 8 | 762 | ||
CR456435.1 | - | 8 | 683 | ||
M16461.1 | - | 7 | 658 | ||
M16462.1 | - | 7 | 596 | ||
Y09501.1 | - | 8 | 696 |