Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133809.1 | NP_598570.1 | 2 | 124 | |
GenBank mRNA | AK129011.1 | - | 137 | ||
AK133074.1 | - | 232 | |||
AK158251.1 | - | 2 | 319 | ||
AK171812.1 | - | 2 | 248 | ||
BC014683.1 | - | 2 | 124 | 4210278 |