Gene Transcripts

Gene Symbol
Wipi1
Entrez Gene ID
52639
Gene Name
WD repeat domain, phosphoinositide interacting 1
Gene Aliases
4930533H01Rik, AW411817, D11Ertd498e
Chromosome Location
Chr. 11: 109573521 - 109611389 on Build GRCm38
UniGene ID
Mm.35817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145940.2 NP_666052.1 2 187
GenBank mRNA AK031672.1 - 2 196
AK034907.1 - 2 231
AK045131.1 - 2 196
AK050495.1 - 2 195
AK159608.1 - 2 216
AK170563.1 - 2 231
BC024811.1 - 2 155
BC024883.1 - 2 151
BC025560.1 - 2 187 5322999

View Full Product Details