Gene Transcripts

Gene Symbol
Wipi1
Entrez Gene ID
52639
Gene Name
WD repeat domain, phosphoinositide interacting 1
Gene Aliases
4930533H01Rik, AW411817, D11Ertd498e
Chromosome Location
Chr. 11: 109573521 - 109611389 on Build GRCm38
UniGene ID
Mm.35817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145940.2 NP_666052.1 7, 8 767
GenBank mRNA AK031672.1 - 7, 8 776
AK034907.1 - 7, 8 811
AK045131.1 - 7, 8 777
AK050495.1 - 7, 8 775
AK159608.1 - 7, 8 796
AK170563.1 - 7, 8 811
BC024811.1 - 7, 8 735
BC024883.1 - 7, 8 731
BC025560.1 - 7, 8 767 5322999

View Full Product Details