Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATRAD17
Entrez Gene ID
836745
Gene Name
RADIATION SENSITIVE 17
Gene Aliases
AT5G66130, K2A18.21, K2A18_21, RAD17, RADIATION SENSITIVE 17
Chromosome Location
Chr.5: 26433964 - 26437222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345748.1 NP_001330043.1 5 - 6 807 67
NM_126011.2 NP_201414.1 5 - 6 807 67
GenBank mRNA AB030250.1 - 5 - 6 707 67

View Full Product Details