Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BET10
Entrez Gene ID
821083
Gene Name
bromodomain and extraterminal domain protein 10
Gene Aliases
AT3G01770, ATBET10, F28J7.10, bromodomain and extraterminal domain protein 10
Chromosome Location
Chr.3: 275333 - 278841 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337357.1 NP_001326912.1 4 - 5 1516 119
NM_111043.3 NP_566151.1 4 - 5 1527 119
GenBank mRNA AY057662.1 - 4 - 5 1465 119
AY099729.1 - 4 - 5 1472 119
AY128906.1 - 3 - 4 1217 119

View Full Product Details