Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPS2
Entrez Gene ID
814882
Gene Name
ureide permease 2
Gene Aliases
ARABIDOPSIS THALIANA UREIDE PERMEASE 2, AT2G03530, ATUPS2, T4M8.3, T4M8_3, ureide permease 2
Chromosome Location
Chr.2: 1071453 - 1074016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084405.2 NP_001077874.1 5 - 6 1302 59
NM_001335188.1 NP_001324121.1 5 - 6 1387 59
NM_001335189.1 NP_001324119.1 5 - 6 1484 59
NM_001335190.1 NP_001324120.1 4 - 5 1561 59
NM_126403.4 NP_178451.2 6 - 7 1310 59

View Full Product Details