Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAK
Entrez Gene ID
830017
Gene Name
HECT ubiquitin protein ligase family protein KAK
Gene Aliases
AT4G38600, AT4G38610, F20M13.160, F20M13_160, KAKTUS, UBIQUITIN-PROTEIN LIGASE 3, UBIQUITIN_PROTEIN LIGASE, UPL3
Chromosome Location
Chr.4: 18040623 - 18049700 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342508.1 NP_001329354.1 10 - 11 5279 88
NM_120021.2 NP_195572.2 10 - 11 4777 88
NM_179236.3 NP_849567.2 10 - 11 5279 88
GenBank mRNA AY265959.1 - 10 - 11 4871 88

View Full Product Details