Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIPS1
Entrez Gene ID
830139
Gene Name
myo-inositol-1-phosphate synthase 1
Gene Aliases
AT4G39800, ATIPS1, ATMIPS1, ATMIPS2, D-myo-Inositol 3-Phosphate Synthase 1, INOSITOL 3-PHOSPHATE SYNTHASE 1, MI-1-P SYNTHASE, MIPS2, MYO-INOSITOL-1-PHOSPHATE SYNTHASE, MYO-INOSITOL-1-PHOSPHATE SYNTHASE 1, MYO-INOSITOL-1-PHOSTPATE SYNTHASE 1, MYO-INOSITOL-1-PHOSTPATE SYNTHASE 2, T19P19.190, T19P19_190, T5J17.3, myo-inositol-1-phosphate synthase 1, myo-inositol-1-phosphate synthase 2
Chromosome Location
Chr.4: 18469255 - 18472048 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120143.4 NP_195690.1 5 - 6 1027 79
GenBank mRNA AF372954.1 - 5 - 6 943 79
AY085931.1 - 5 - 6 943 79
BT001931.1 - 5 - 6 872 79

View Full Product Details