Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN1A
Entrez Gene ID
816580
Gene Name
26S proteasome regulatory subunit S2 1A
Gene Aliases
26S PROTEASOME REGULATORY SUBUNIT S2 1A, 26S proteasome regulatory subunit S2 1A, AT2G20580, ATRPN1A, F23N11.10, F23N11_10
Chromosome Location
Chr.2: 8858962 - 8864865 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127618.4 NP_565477.1 21 - 22 2670 71
GenBank mRNA AF360216.1 - 21 - 22 2622 71
AY040061.1 - 21 - 22 2421 71
AY230828.1 - 21 - 22 2505 71

View Full Product Details