Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAPT2
Entrez Gene ID
825529
Gene Name
aminoalcoholphosphotransferase
Gene Aliases
AMINOALCOHOLPHOSPHOTRANSFERASE, AT3G25585, ATAAPT2, aminoalcoholphosphotransferase
Chromosome Location
Chr.3: 9295175 - 9298746 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035689.2 NP_001030766.1 8 - 9 1003 130
NM_001035690.1 NP_001030767.1 9 - 10 1062 130
NM_001338758.1 NP_001327446.1 8 - 9 1167 130
NM_001338759.1 NP_001327445.1 7 - 8 612 130
NM_113456.4 NP_189186.1 9 - 10 1066 130
NM_180413.2 NP_850744.1 9 - 10 865 130
GenBank mRNA AF091844.1 - 8 - 9 705 130
AF360151.1 - 9 - 10 930 130
AY054269.1 - 9 - 10 914 130
AY142609.1 - 8 - 9 702 130

View Full Product Details