Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD6
Entrez Gene ID
824538
Gene Name
ARF-GAP domain 6
Gene Aliases
ARF-GAP domain 6, AT3G53710
Chromosome Location
Chr.3: 19903234 - 19905590 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035777.2 NP_001030854.1 1 - 2 776 81
NM_001339627.1 NP_001319740.1 1 - 2 860 81
NM_115231.4 NP_190939.1 1 - 2 935 81
GenBank mRNA BT030393.1 - 1 - 2 764 81

View Full Product Details