Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACC2
Entrez Gene ID
840522
Gene Name
acetyl-CoA carboxylase 2
Gene Aliases
AT1G36180, F15C21.2, F15C21_2, acetyl-CoA carboxylase 2
Chromosome Location
Chr.1: 13545965 - 13558600 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103314.5 NP_174850.4 31 - 32 7087 65

View Full Product Details