The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085110.1 | NP_001078579.1 | 1 - 2 | 150 | 108 |
NM_121374.4 | NP_196875.1 | 2 - 3 | 434 | 108 | |
GenBank mRNA | AF090372.1 | - | 2 - 3 | 101 | 108 |
AF195648.1 | - | 2 - 3 | 112 | 108 | |
AY120716.1 | - | 2 - 3 | 126 | 108 | |
BT000058.1 | - | 1 - 2 | 59 | 108 |