Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SK13
Entrez Gene ID
831316
Gene Name
shaggy-like kinase 13
Gene Aliases
AT5G14640, ATSK13, SHAGGY-LIKE KINASE 13, T15N1.130, T15N1_130, shaggy-like kinase 13
Chromosome Location
Chr.5: 4718994 - 4722371 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343350.1 NP_001329975.1 11 - 12 1146 95
NM_121468.4 NP_196968.2 11 - 12 1350 95
GenBank mRNA AY065043.1 - 11 - 12 1258 95
BT010466.1 - 10 - 11 1045 95

View Full Product Details