Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DRM1
- Entrez Gene ID
-
831390
- Gene Name
- domains rearranged methylase 1
- Gene Aliases
-
AT5G15380, F8M21.270, F8M21_270, domains rearranged methylase 1
- Chromosome Location
-
Chr.5: 4991347 - 4994924 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121542.3 |
NP_197042.2 |
7 - 8 |
644 |
79 |
View Full Product Details