Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CSA1
- Entrez Gene ID
-
831656
- Gene Name
- disease resistance protein (TIR-NBS-LRR class)
- Gene Aliases
-
AT5G17880, MPI7.20, MPI7_20, constitutive shade-avoidance1
- Chromosome Location
-
Chr.5: 5908834 - 5913317 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121794.2 |
NP_197290.1 |
5 - 6 |
3535 |
72 |
View Full Product Details