Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ORP1B
- Entrez Gene ID
-
817654
- Gene Name
- OSBP(oxysterol binding protein)-related protein 1B
- Gene Aliases
-
AT2G31030, F7F1.24, F7F1_24, OSBP(oxysterol binding protein)-related protein 1B
- Chromosome Location
-
Chr.2: 13206004 - 13207815 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128657.1 |
NP_180660.1 |
1 - 2 |
241 |
79 |
View Full Product Details