Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP26
Entrez Gene ID
817867
Gene Name
receptor like protein 26
Gene Aliases
AT2G33050, AtRLP26, F25I18.21, F25I18_21, receptor like protein 26
Chromosome Location
Chr.2: 14021728 - 14024398 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128865.2 NP_180864.1 1 - 1 1045 132
GenBank mRNA BT002326.1 - 1 - 1 1045 132

View Full Product Details