Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACS1
Entrez Gene ID
825324
Gene Name
ACC synthase 1
Gene Aliases
1-AMINOCYCLOPROPANE-1-CARBOXYLATE SYNTHASE 1, ACC synthase 1, ARABIDOPSIS THALIANA 1-AMINOCYCLOPROPANE-1-CARBOXYLATE SYNTHASE 1, AT-ACS1, AT3G61510
Chromosome Location
Chr.3: 22763495 - 22766010 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116016.2 NP_191710.1 3 - 4 739 71
GenBank mRNA AY133715.1 - 3 - 4 459 71
U26543.1 - 3 - 4 459 71

View Full Product Details