Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PDIL1-5
- Entrez Gene ID
-
841656
- Gene Name
- PDI-like 1-5
- Gene Aliases
-
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 3, AT1G52260, ATPDI3, ATPDIL1-5, F19K6.17, F19K6_17, PDI-like 1-5, PDI3, PROTEIN DISULFIDE ISOMERASE 3
- Chromosome Location
-
Chr.1: 19460482 - 19463477 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104105.3 |
NP_175636.2 |
2 - 3 |
858 |
153 |
GenBank mRNA |
BT030322.1 |
- |
2 - 3 |
646 |
153 |
View Full Product Details