Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPC1
Entrez Gene ID
841765
Gene Name
phosphoenolpyruvate carboxylase 1
Gene Aliases
AT1G53310, ATPEPC1, ATPPC1, F12M16.21, F12M16_21, PEP(PHOSPHOENOLPYRUVATE) CARBOXYLASE 1, PEPC1, phosphoenolpyruvate carboxylase 1
Chromosome Location
Chr.1: 19883935 - 19888413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036101.2 NP_001031178.1 8 - 9 1353 104
NM_001036102.3 NP_001031179.1 7 - 8 1484 104
NM_104209.3 NP_175738.1 8 - 9 1414 104
GenBank mRNA AJ532901.1 - 7 - 8 1226 104
AY057507.1 - 8 - 9 1292 104
BT000647.1 - 7 - 8 1226 104

View Full Product Details