The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001333882.1 | NP_001322705.1 | 2 - 3 | 3002 | 71 |
NM_001333883.1 | NP_001322704.1 | 2 - 3 | 2679 | 71 | |
NM_001333884.1 | NP_001322706.1 | 3 - 4 | 2824 | 71 | |
NM_001333885.1 | NP_001322703.1 | 3 - 4 | 2407 | 71 | |
NM_001333886.1 | NP_001322702.1 | 2 - 3 | 2321 | 71 | |
NM_104705.4 | NP_176221.1 | 2 - 3 | 2679 | 71 | |
GenBank mRNA | AF155151.1 | - | 1 - 2 | 1993 | 71 |
KJ138959.1 | - | 1 - 2 | 1993 | 71 |