Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS10
Entrez Gene ID
842309
Gene Name
trehalose phosphate synthase
Gene Aliases
AT1G60140, ATTPS10, T13D8.4, T13D8_4, TREHALOSE PHOSPHATE SYNTHASE 10, trehalose phosphate synthase
Chromosome Location
Chr.1: 22176934 - 22181740 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333882.1 NP_001322705.1 3 - 4 3265 58
NM_001333884.1 NP_001322706.1 4 - 5 3087 58
NM_001333885.1 NP_001322703.1 4 - 5 2670 58
NM_001333886.1 NP_001322702.1 3 - 4 2584 58
NM_104705.4 NP_176221.1 3 - 4 2942 58
GenBank mRNA AF155151.1 - 2 - 3 2256 58
KJ138959.1 - 2 - 3 2256 58

View Full Product Details