Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRA3
Entrez Gene ID
842869
Gene Name
Endonuclease/exonuclease/phosphatase family protein
Gene Aliases
AT1G65580, F5I14.11, F5I14_11, FRAGILE FIBER3
Chromosome Location
Chr.1: 24377263 - 24384086 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105232.3 NP_176736.2 1 - 2 863 71
GenBank mRNA AY761186.1 - 1 - 2 836 71

View Full Product Details