Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AOAT2
Entrez Gene ID
843395
Gene Name
alanine-2-oxoglutarate aminotransferase 2
Gene Aliases
AT1G70580, F5A18.24, F5A18_24, GGT2, GLUTAMATE:GLYOXYLATE AMINOTRANSFERASE 2, alanine-2-oxoglutarate aminotransferase 2
Chromosome Location
Chr.1: 26612359 - 26616071 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036185.1 NP_001031262.1 3 - 4 332 113
NM_001036186.2 NP_001031263.1 3 - 4 444 113
NM_105726.3 NP_177215.1 3 - 4 670 113
NM_202393.2 NP_974122.1 3 - 4 639 113
GenBank mRNA AF479640.1 - 2 - 3 204 113
AK316788.1 - 3 - 4 271 113
AY035130.1 - 3 - 4 310 113
AY062982.1 - 2 - 3 185 113

View Full Product Details