Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC3
Entrez Gene ID
819581
Gene Name
plastid transcriptionally active 3
Gene Aliases
AT3G04260, PDE324, PIGMENT DEFECTIVE 324, T6K12.12, T6K12_12, plastid transcriptionally active 3
Chromosome Location
Chr.3: 1122948 - 1127581 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111297.4 NP_187076.2 10 - 11 2266 113

View Full Product Details