Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PTAC3
- Entrez Gene ID
-
819581
- Gene Name
- plastid transcriptionally active 3
- Gene Aliases
-
AT3G04260, PDE324, PIGMENT DEFECTIVE 324, T6K12.12, T6K12_12, plastid transcriptionally active 3
- Chromosome Location
-
Chr.3: 1122948 - 1127581 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111297.4 |
NP_187076.2 |
6 - 7 |
1046 |
63 |
View Full Product Details