Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX4
Entrez Gene ID
823624
Gene Name
cation/H+ exchanger 4
Gene Aliases
AT3G44900, ATCHX4, cation/H+ exchanger 4
Chromosome Location
Chr.3: 16388724 - 16391582 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114359.2 NP_190076.1 2 - 3 1319 93
GenBank mRNA DQ499020.1 - 2 - 3 1319 93

View Full Product Details