Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPI1
Entrez Gene ID
828859
Gene Name
proton pump interactor 1
Gene Aliases
AT4G27500, F27G19.100, F27G19_100, proton pump interactor 1
Chromosome Location
Chr.4: 13743011 - 13746211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341859.1 NP_001328990.1 2 - 3 557 70
NM_118885.4 NP_194480.2 2 - 3 647 70
GenBank mRNA AJ002020.2 - 2 - 3 466 70
AY050365.1 - 2 - 3 486 70
AY143914.1 - 1 - 2 361 70

View Full Product Details