Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL3
Entrez Gene ID
829788
Gene Name
beta-galactosidase 3
Gene Aliases
AT4G36360, F23E13.200, F23E13_200, beta-galactosidase 3
Chromosome Location
Chr.4: 17176342 - 17181466 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119799.4 NP_568001.1 4 - 5 781 93
NM_179175.2 NP_849506.1 4 - 5 576 93
GenBank mRNA AJ270299.1 - 4 - 5 498 93
AY056285.1 - 4 - 5 557 93
AY091432.1 - 4 - 5 458 93

View Full Product Details