Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOM
Entrez Gene ID
837322
Gene Name
ATP-dependent helicase family protein
Gene Aliases
AT1G08060, MAINTENANCE OF METHYLATION, MOM1, MORPHEUS MOLECULE, MORPHEUS MOLECULE 1, T6D22.14, T6D22_14
Chromosome Location
Chr.1: 2501704 - 2511084 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331755.1 NP_001322007.1 13 - 14 5378 62
NM_100680.4 NP_563806.1 13 - 14 5375 62
NM_179277.1 NP_849608.1 12 - 13 5107 62

View Full Product Details