Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLUR2
Entrez Gene ID
829682
Gene Name
glutamate receptor 2
Gene Aliases
AT4G35290, ATGLR3.2, ATGLUR2, F23E12.150, F23E12_150, GLR3.2, GLUTAMATE RECEPTOR 3.2, glutamate receptor 2
Chromosome Location
Chr.4: 16789531 - 16793685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342335.1 NP_001320141.1 5 - 6 2441 63
NM_001342336.1 NP_001329586.1 6 - 7 2235 63
NM_001342337.1 NP_001329588.1 6 - 7 2145 63
NM_001342338.1 NP_001329587.1 6 - 7 2161 63
NM_119695.4 NP_567981.1 5 - 6 2067 63
NM_202957.3 NP_974686.1 6 - 7 2235 63

View Full Product Details