Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEA1
Entrez Gene ID
837332
Gene Name
K+ efflux antiporter 1
Gene Aliases
AT1G01790, ATKEA1, K+ EFFLUX ANTIPORTER 1, K+ efflux antiporter 1, T1N6.21, T1N6_21
Chromosome Location
Chr.1: 284350 - 291203 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331307.1 NP_001320631.1 17 - 18 3650 95
NM_100062.5 NP_171684.2 18 - 19 3674 95

View Full Product Details