Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC1A
Entrez Gene ID
839288
Gene Name
secretory 1A
Gene Aliases
AT1G02010, secretory 1A
Chromosome Location
Chr.1: 347613 - 352545 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197959.1 NP_001184888.1 13 - 14 1199 89
NM_001331318.1 NP_001320381.1 13 - 14 1368 89
NM_001331319.1 NP_001320383.1 12 - 13 1439 89
NM_001331320.1 NP_001320382.1 12 - 13 1433 89
NM_001331321.1 NP_001320384.1 7 - 8 727 89
NM_100081.5 NP_563643.4 13 - 14 1560 89

View Full Product Details