Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP5
Entrez Gene ID
837588
Gene Name
zinc finger protein 5
Gene Aliases
AT1G10480, T10O24.9, T10O24_9, zinc finger protein 5
Chromosome Location
Chr.1: 3449567 - 3451226 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100922.3 NP_172518.1 1 - 1 1211 169

View Full Product Details