Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOT3
Entrez Gene ID
830889
Gene Name
Phototropic-responsive NPH3 family protein
Gene Aliases
AT5G10250, DEFECTIVELY ORGANIZED TRIBUTARIES 3, F18D22.20, F18D22_20
Chromosome Location
Chr.5: 3216881 - 3219436 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343097.1 NP_001330410.1 2 - 3 267 147
NM_121063.2 NP_196587.1 2 - 3 259 147

View Full Product Details