Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP15
Entrez Gene ID
831046
Gene Name
arabinogalactan protein 15
Gene Aliases
ARABINOGALACTAN PROTEIN 15, AT5G11740, ATAGP15, T22P22.130, T22P22_130, arabinogalactan protein 15
Chromosome Location
Chr.5: 3783930 - 3784963 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121212.3 NP_196735.1 1 - 1 660 103
GenBank mRNA AF375418.1 - 1 - 1 354 103

View Full Product Details