Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIL2
Entrez Gene ID
817254
Gene Name
DNA polymerase epsilon catalytic subunit
Gene Aliases
AT2G27120, POL2B, T20P8.17, T20P8_17, TILTED 2
Chromosome Location
Chr.2: 11581214 - 11594397 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336102.1 NP_001325161.1 34 - 35 4493 71
NM_001336103.1 NP_001325160.1 34 - 35 4496 71
NM_128270.3 NP_180280.2 34 - 35 4493 71

View Full Product Details