Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACA2
Entrez Gene ID
817367
Gene Name
alpha carbonic anhydrase 2
Gene Aliases
AT2G28210, ATACA2, T3B23.12, T3B23_12, alpha carbonic anhydrase 2
Chromosome Location
Chr.2: 12029666 - 12033096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336148.1 NP_001318303.1 3 - 4 563 80

View Full Product Details