Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD5
Entrez Gene ID
821783
Gene Name
IQ-domain 5
Gene Aliases
AT3G22190, IQ-domain 5
Chromosome Location
Chr.3: 7830905 - 7834135 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203017.2 NP_001189946.1 3 - 4 699 60
NM_001338571.1 NP_001319617.1 3 - 4 761 60
NM_001338572.1 NP_001325926.1 2 - 3 927 60
NM_113116.4 NP_188858.4 3 - 4 460 60

View Full Product Details