Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAG1
Entrez Gene ID
822140
Gene Name
TRAM, LAG1 and CLN8 (TLC) lipid-sensing domain containing protein
Gene Aliases
AT3G25540, LAG One Homologue 1, LAG1 HOMOLOG 1, LOH1, LONGEVITY ASSURANCE GENE 1
Chromosome Location
Chr.3: 9274335 - 9276685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338755.1 NP_001319643.1 3 - 4 851 105
NM_001338756.1 NP_001327930.1 4 - 5 855 105
NM_113450.5 NP_566769.1 4 - 5 855 105
GenBank mRNA AF198179.1 - 3 - 4 580 105
AF360152.1 - 3 - 4 686 105
AY142597.1 - 3 - 4 530 105

View Full Product Details