Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATHAL3B
- Entrez Gene ID
-
2745818
- Gene Name
- Flavoprotein
- Gene Aliases
-
AT1G48605, HAL3, HAL3B, HALOTOLERANCE DETERMINANT 3, HALOTOLERANCE DETERMINANT 3B
- Chromosome Location
-
Chr.1: 17969395 - 17970551 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_202265.3 |
NP_973994.1 |
2 - 3 |
617 |
121 |
View Full Product Details