The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198045.1 | NP_001184974.1 | 6 - 7 | 1292 | 95 |
NM_101144.5 | NP_172734.2 | 6 - 7 | 1347 | 95 | |
GenBank mRNA | AK176324.1 | - | 6 - 7 | 1272 | 95 |
BT010888.1 | - | 6 - 7 | 1094 | 95 |