Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bZIP44
Entrez Gene ID
843875
Gene Name
basic leucine-zipper 44
Gene Aliases
AT1G75390, AtbZIP44, F1B16.8, F1B16_8, basic leucine-zipper 44
Chromosome Location
Chr.1: 28291698 - 28293008 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084357.2 NP_001077826.1 2 - 2 950 134
NM_106193.4 NP_177672.2 1 - 1 1020 134
GenBank mRNA AK318829.1 - 2 - 2 816 134
AK318991.1 - 2 - 2 764 134

View Full Product Details