Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECT8
Entrez Gene ID
844267
Gene Name
evolutionarily conserved C-terminal region 8
Gene Aliases
AT1G79270, evolutionarily conserved C-terminal region 8
Chromosome Location
Chr.1: 29815470 - 29819678 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334878.1 NP_001321640.1 5 - 6 1629 97
NM_001334879.1 NP_001321639.1 5 - 6 1624 97
NM_001334880.1 NP_001321641.1 5 - 6 987 97
NM_106577.4 NP_565205.1 5 - 6 1340 97
GenBank mRNA AF326910.1 - 5 - 6 934 97
AY113874.1 - 5 - 6 653 97

View Full Product Details