Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HP6
Entrez Gene ID
844350
Gene Name
histidine phosphotransfer protein 6
Gene Aliases
AHP6, AT1G80100, F18B13.18, F18B13_18, histidine phosphotransfer protein 6
Chromosome Location
Chr.1: 30133718 - 30134937 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334957.1 NP_001321526.1 1 - 2 296 97
NM_106659.3 NP_178127.2 1 - 2 292 97
GenBank mRNA DQ093643.1 - 1 - 2 288 97

View Full Product Details