Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDP6
Entrez Gene ID
820199
Gene Name
FAD-dependent oxidoreductase family protein
Gene Aliases
AT3G10370, SUGAR-DEPENDENT 6
Chromosome Location
Chr.3: 3216339 - 3219270 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111872.6 NP_187648.1 3 - 4 1332 66
GenBank mRNA AY063863.1 - 3 - 4 1257 66
AY096363.1 - 3 - 4 1169 66

View Full Product Details