Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ABCB19
- Entrez Gene ID
-
822519
- Gene Name
- ATP binding cassette subfamily B19
- Gene Aliases
-
AT3G28860, ATABCB19, ATMDR1, ATMDR11, ATP BINDING CASSETTE SUBFAMILY B19, ATP binding cassette subfamily B19, ATP-binding cassette B19, ATPGP19, Arabidopsis thaliana ATP-binding cassette B19, MDR1, MDR11, MULTIDRUG RESISTANCE PROTEIN 11, P-GLYCOPROTEIN 19, PGP19
- Chromosome Location
-
Chr.3: 10870005 - 10877578 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113807.3 |
NP_189528.1 |
2 - 3 |
602 |
74 |
View Full Product Details